Merck
  • Ca2+/calmodulin-dependent protein kinase IIγ enhances stem-like traits and tumorigenicity of lung cancer cells.
Ca2+/calmodulin-dependent protein kinase IIγ enhances stem-like traits and tumorigenicity of lung cancer cells.
Oncotarget (2015-05-13)
Shoujie Chai, Xia Xu, Yongfang Wang, You Zhou, Chenchen Zhang, Yiming Yang, Ying Yang, Haiyan Xu, Rongzhen Xu, Kai Wang
ZUSAMMENFASSUNG
Highly tumorigenic stem-like cells, considered tumor-initiating cells (TICs), are the main cause of lung cancer initiation, relapse, and drug resistance. In this study, we identified that Ca2+/calmodulin-dependent protein kinase IIγ (CaMKIIγ) was aberrantly expressed in highly tumorigenic stem-like lung cancer cells, and was also correlated with poor prognosis in human lung cancer. Functionally, CaMKIIγ enhanced stem-like traits and the tumorigenicity of lung cancer cells in an Akt- and β-catenin-dependent manner. In addition, we found that CaMKIIγ upregulated Oct4 expression via Akt-mediated histone acetylation. Taken together, our findings reveal a critical role of CaMKIIγ in regulating the stemness and tumorigenicity of lung cancer cells and offer a promising therapeutic target for TICs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Akt Inhibitor IV, The Akt Inhibitor IV, also referenced under CAS 681281-88-9, controls the biological activity of Akt. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.
Sigma-Aldrich
Wnt-Signalweg-Inhibitor XI, CCT036477 – CAS 305372-78-5 – Calbiochem, The Wnt Pathway Inhibitor XI, CCT036477, also referenced under CAS 305372-78-5, controls the biological activity of Wnt Pathway. This small molecule/inhibitor is primarily used for Cancer applications.