Merck
  • Prenatal findings and the genetic diagnosis of fetal overgrowth disorders: Simpson-Golabi-Behmel syndrome, Sotos syndrome, and Beckwith-Wiedemann syndrome.
Prenatal findings and the genetic diagnosis of fetal overgrowth disorders: Simpson-Golabi-Behmel syndrome, Sotos syndrome, and Beckwith-Wiedemann syndrome.
Taiwanese journal of obstetrics & gynecology (2012-07-17)
Chih-Ping Chen
ZUSAMMENFASSUNG
With the advent of prenatal sonography, fetal overgrowth can be easily detected. Prenatal-onset overgrowth can be secondary to normal variants of familial tall stature, familial rapid maturation, diabetic macrosomia, and congenital nesidioblastosis, or prenatal-onset overgrowth can be primary due to pathological overgrowth disorders. This article provides a comprehensive review of the prenatal findings and the genetic diagnosis of some of the pathological prenatal-onset overgrowth disorders, such as Simpson-Golabi-Behmel syndrome, Sotos syndrome, and Beckwith-Wiedemann syndrome.