Merck
  • Antiinflammatory and antinociceptive effects of 1,8-cineole a terpenoid oxide present in many plant essential oils.
Antiinflammatory and antinociceptive effects of 1,8-cineole a terpenoid oxide present in many plant essential oils.
Phytotherapy Research (2000)
Santos FA, Rao VS.