Merck
  • Investigation of the clinical significance of the growth hormone-releasing peptide-2 test for the diagnosis of secondary adrenal failure.
Investigation of the clinical significance of the growth hormone-releasing peptide-2 test for the diagnosis of secondary adrenal failure.
Endocrine journal (2016-03-30)
Hiroshi Arimura, Hiroshi Hashiguchi, Kiyoaki Yamamoto, Atsushi Shinnakasu, Aiko Arimura, Akira Kikuchi, Takahisa Deguchi, Mika Habu, Singo Fujio, Kazunori Arita, Yoshihiko Nishio
ZUSAMMENFASSUNG
The aim of this study was to evaluate the ability of the growth hormone-releasing peptide-2 (GHRP-2) test to clinically diagnose hypothalamo-pituitary-adrenal (HPA) axis failure. We performed an insulin tolerance test (ITT), CRH stimulation test, and GHRP-2 test on 47 patients suspected of having a hypothalamo-pituitary disorder. Patients with pituitary disorders had significantly lower ACTH responses to the GHRP-2 test compared to patients with hypothalamic disorders and the control group. In contrast, peak cortisol levels in response to the GHRP-2 test were significantly lower in both hypothalamic and pituitary disorder cases compared with the control group. Assignment of a cut-off value of 11.6 μg/dL for the peak serum cortisol level demonstrated that the GHRP-2 test was able to predict secondary hypoadrenalism with 88.9% specificity and 89.7% sensitivity. The responses of ACTH and cortisol to the GHRP-2 test had no correlation to the CRH test, suggesting the involvement of a different mechanism of ACTH secretion. These results indicate that the GHRP-2 test may induce ACTH secretion from the pituitary gland through direct stimulation. Although the GHRP-2 test does not have the same predictive value as the insulin tolerance test (ITT), it has similar diagnostic potential as the CRH stimulation test for evaluating HPA axis failure.