Merck
  • Toward Système International d'Unité-traceable protein quantification: from amino acids to proteins.
Toward Système International d'Unité-traceable protein quantification: from amino acids to proteins.
Analytical biochemistry (2008-03-14)
William I Burkitt, Caroline Pritchard, Cristian Arsene, André Henrion, David Bunk, Gavin O'Connor
ZUSAMMENFASSUNG
Here we present a demonstration of the proof of principle that absolute concentration of a protein within a mixture of other proteins can be measured with SI traceability. The method used was based on tryptic digestion of a protein followed by quantification using double exact matching isotope dilution mass spectrometry (IDMS) of the peptides released. To provide full SI traceability to measurements of protein concentration we demonstrated a method of SI traceable peptide quantification in which the peptide standards used were quantified by an amino acid analysis method that incorporated double exact matching IDMS and amino acid standards of known purity. The concentration of the protein was therefore determined based upon the concentration of tryptic peptides, which in turn had been quantified based upon amino acid standards. This allowed fully SI-traceable measurements of protein concentration to be made. Important caveats in the implementation of this approach are also discussed and examples of how these can have detrimental effects on the measurements are shown.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Alcohol-Dehydrogenase aus Saccharomyces cerevisiae, ≥300 units/mg protein, lyophilized powder (contains buffer salts), Mw 141-151 kDa
Sigma-Aldrich
Angiotensin I human, ≥90% (HPLC), powder, suitable for mammalian cell culture
Sigma-Aldrich
Insulin-Kette-B-oxidiert aus Rinderpankreas, ≥80% (HPLC), powder