Merck
  • 3-Nitrofluoranthene (3-NF) but not 3-aminofluoranthene (3-AF) elicits apoptosis as well as programmed necrosis in Hepa1c1c7 cells.
3-Nitrofluoranthene (3-NF) but not 3-aminofluoranthene (3-AF) elicits apoptosis as well as programmed necrosis in Hepa1c1c7 cells.
Toxicology (2008-12-02)
Nana Asare, Marit Låg, Dominique Lagadic-Gossmann, Mary Rissel, Per Schwarze, Jørn A Holme
ZUSAMMENFASSUNG
In this study, we show that the environmental pollutant, 3-nitrofluoranthene (3-NF) but not its amine form, 3-aminofluoranthene (3-AF), induces apoptosis as well as regulated necrosis with necroptotic features in Hepa1c1c7 cells. Upon exposure to 3-NF, both typical apoptotic and necrotic cells were observed. A large number of the cells exhibited a characteristic partial nuclear chromatin condensation. Cycloheximide completely attenuated 3-NF-induced cell death. Activation of caspase-8, -9, and -3 were observed. Moreover, Z-VAD-FMK decreased the apoptotic cells, whereas the number of propidium iodide (PI)-positive cells with partial chromatin condensation was reduced by Nec-1, an inhibitor of receptor interacting protein (RIP-1). Cyp1a1, but not nitric oxide synthase (NOS), appears to be involved in activation of 3-NF to reactive metabolites. Increase in the number as well as size of lysosomes, myelinosomes, and activation of autophagy were also observed. 3-NF induced phosphorylation of ERK1/2, JNK and p38 MAPKs. Interestingly, while inhibitors of ERK1/2 and JNK reduced apoptotic as well as necrotic cell death, the p38 inhibitor, SB202190 reduced only the necrotic cell death. Taken together, 3-NF elicits both apoptosis and a caspase-independent programmed cell death (PCD) with autophagic characteristics. Conversely, with 3-AF, no apparent cytotoxic effects besides a reduction in cell proliferation was observed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ribonuklease A aus Rinderpankreas, Type II-A, ≥60% (SDS-PAGE), >= 60 Kunitz units/mg protein
Sigma-Aldrich
3-Nitrofluoranthen, technical grade, 90%
3-Nitrofluoranthen, BCR®, certified reference material