Merck
  • Determination of aflatoxin M1 in milk samples by means of an inductively coupled plasma mass spectrometry-based immunoassay.
Determination of aflatoxin M1 in milk samples by means of an inductively coupled plasma mass spectrometry-based immunoassay.
Food chemistry (2017-04-15)
Emma Pérez, Pascual Martínez-Peinado, Francisco Marco, Luis Gras, José Miguel Sempere, Juan Mora, Guillermo Grindlay
ZUSAMMENFASSUNG
An inductively coupled plasma mass spectrometry (ICP-MS)-based immunoassay has been developed to quantify aflatoxin M1 (AFM1) at ultra-trace levels in milk samples. AFM1 detection is carried out by means of a competitive immunoassay using secondary biotinylated antibodies and streptavidin-conjugated Au nanoparticles. After acid addition, nanoparticles are decomposed and Au signal is registered by means of ICP-MS. Results demonstrate that, under optimum conditions, the limit of detection of the immunoassay (0.005μgkg