Merck
Bone regeneration enhancement by ultra-violet (UV) treatment for uHA/PLLA absorbable mesh.
Journal of cranio-maxillo-facial surgery : official publication of the European Association for Cranio-Maxillo-Facial Surgery (2017-03-23)
Hiroumi Ikawa, Akinori Moroi, Kunio Yoshizawa, Yuriko Saida, Asami Hotta, Takamitsu Tsutsui, Kenichi Fukaya, Ryota Hiraide, Akihiro Takayama, Tatsuya Tsunoda, Yuki Saito, Koichiro Ueki
ZUSAMMENFASSUNG
The purpose of this study was to evaluate the effect of bone regeneration enhancement by ultra-violet (UV) treatment of an unsintered hydroxyapatite (u-HA)/poly-l-lactic acid (PLLA) mesh in rabbitnasal bone. Thirty adult male Japanese white rabbits (12-16 weeks, 2.5-3.0 kg) were used in this study. After incising along the nasal bone, 6 × 6 × depth 1 mm two bone defects were made on both sides. In the UV group (n = 30), the defects were covered with uHA/PLLA mesh, treated by UV (wavelength 172 nm, tube wall illumination 13 mW/cm The UV group had a significantly higher number of ALP stained cells than the control group after 1, 2, and 3 weeks (P < 0.05). The bone ratio was also significantly higher in the UV group than in the control group after 1, 2, 3, and 4 weeks (P < 0.05). This study suggests that bone regeneration can be enhanced by UV treatment using an uHA/PLLA mesh.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)