- The parathyroid hormone family member TIP39 interacts with sarco/endoplasmic reticulum Ca
The parathyroid hormone family member TIP39 interacts with sarco/endoplasmic reticulum Ca
Experimental dermatology (2017-01-18)
Emi Sato, Michael R Williams, James A Sanford, George L Sen, Takekuni Nakama, Shinichi Imafuku, Richard L Gallo
PMID28094886
ZUSAMMENFASSUNG
Darier disease (DD) is a genetic skin disease that is associated with mutations in the ATP2A2 gene encoding the type 2 sarco/endoplasmic reticulum (ER) Ca
MATERIALIEN