Merck
Structures and activities of protease inhibitors of microbial origin.
Methods in enzymology (1976-01-01)
H Umezawa
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Chymostatin, microbial
Sigma-Aldrich
Antipain -dihydrochlorid aus microbieller Quelle, protease inhibitor
Sigma-Aldrich
Phosphoramidon Dinatriumsalz, ≥97% (HPLC)
Sigma-Aldrich
Leupeptin -hemisulfat (Salz), >4000 U/mg
Sigma-Aldrich
Pepstatin, water soluble, semisynthetic, >=95% pure