Merck
  • Optimization of a Luminescence-Based High-Throughput Screening Assay for Detecting Apyrase Activity.
Optimization of a Luminescence-Based High-Throughput Screening Assay for Detecting Apyrase Activity.
SLAS discovery : advancing life sciences R & D (2016-11-09)
John R Veloria, Ashwini K Devkota, Eun Jeong Cho, Kevin N Dalby
ZUSAMMENFASSUNG
Apyrase is a calcium-activated enzyme that catalyzes the conversion of adenosine triphosphate (ATP) to adenosine diphosphate (ADP), adenosine monophosphate (AMP), and P
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Apyrase aus Kartoffeln, ATPase ≥200 units/mg protein, lyophilized powder