Merck
  • Tribbles Pseudokinase 3 Induces Both Apoptosis and Autophagy in Amyloid-β-induced Neuronal Death.
Tribbles Pseudokinase 3 Induces Both Apoptosis and Autophagy in Amyloid-β-induced Neuronal Death.
The Journal of biological chemistry (2016-12-25)
Suraiya Saleem, Subhas Chandra Biswas
ZUSAMMENFASSUNG
Amyloid-β (Aβ)-induced neuron death is considered central to the pathogenesis of Alzheimer's disease (AD). Among several death modalities, autophagy and apoptosis play important roles in Aβ-induced neuron death suggesting that there may be regulatory mechanisms that initiate both cell death pathways. However, molecules that govern both pathways have not been identified. Here, we report that, upon Aβ treatment, tribbles pseudokinase 3 (Trib3, an ortholog of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3-Methyladenin, autophagy inhibitor