Merck
  • A novel synaptic junction preparation for the identification and characterization of cleft proteins.
A novel synaptic junction preparation for the identification and characterization of cleft proteins.
PloS one (2017-04-01)
Amelia Burch, Jung-Hwa Tao-Cheng, Ayse Dosemeci
ZUSAMMENFASSUNG
Identification of synaptic cleft components has been hampered by the lack of a suitable preparation enriched in synaptic junctions devoid of adjoining peripheral membranes. Prior strategies for the isolation of synaptic junctions, relying on detergents for the removal of peripheral membranes, resulted in substantial loss of membranes lining the cleft. Here, a novel, detergent-free method is described for the preparation of a synaptic junction (SJ) fraction, using phospholipase A2. Limited digestion of synaptic plasma membrane (SPM) fraction with phospholipase A2 followed by centrifugation over a sucrose cushion results in selective removal of membranes peripheral to the cleft while junctional membranes remain relatively intact as observed by electron microscopy. Enrichment in synaptic junctional structures and loss of membranes peripheral to the junctional area are further verified by demonstrating enrichment in PSD-95 and loss in mGluR5, respectively. The SJ fraction is enriched in neuroligins and neurexins, in agreement with immuno-electron microscopy data showing their selective localization to the junctional area. Among additional cell adhesion molecules tested, N-cadherin and specific isoforms of the SynCAM and SALM families also show marked enrichment in the SJ fraction, suggesting preferential localization at the synaptic cleft while others show little enrichment or decrease, suggesting that they are not restricted to or concentrated at the synaptic cleft. Treatment of the SJ fraction with glycosidases results in electrophoretic mobility shifts of all cell adhesion molecules tested, indicating glycosylation at the synaptic cleft. Biochemical and ultrastructural data presented indicate that the novel synaptic junction preparation can be used as a predictive tool for the identification and characterization of the components of the synaptic cleft.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phospholipase A2 aus Honigbienengift (Apis mellifera), salt-free, lyophilized powder, 600-2400 units/mg protein
Sigma-Aldrich
Enzymatisches Protein-Deglykolysierungs-Kit
Sigma-Aldrich
Anti-metabotroper-Glutamatrezeptor-5-Antikörper, Schmerz, Chemicon®, from rabbit