Merck
  • Quantitative in vivo imaging of tissue factor expression in glioma using dynamic contrast-enhanced MRI derived parameters.
Quantitative in vivo imaging of tissue factor expression in glioma using dynamic contrast-enhanced MRI derived parameters.
European journal of radiology (2017-07-03)
Xiao Chen, Tian Xie, Jingqin Fang, Wei Xue, Haipeng Tong, Houyi Kang, Sumei Wang, Yizeng Yang, Minhui Xu, Weiguo Zhang
ZUSAMMENFASSUNG
Tissue Factor (TF) has been well established in angiogenesis, invasion, metastasis, and prognosis in glioma. A noninvasive assessment of TF expression status in glioma is therefore of obvious clinical relevance. Dynamic contrast-enhanced (DCE) MRI parameters have been used to evaluate microvascular characteristics and predict molecular expression status in tumors. Our aim is to investigate whether quantitative DCE-MRI parameters could assess TF expression in glioma. Thirty-two patients with histopathologically diagnosed supratentorial glioma who underwent DCE-MRI were retrospectively recruited. Extended Tofts linear model was used for DCE-MRI post-processing. Hot-spot, whole tumor cross-sectional approaches, and histogram were used for analysis of model based parameters. Four serial paraffin sections of each case were stained with TF, CD105, CD34 and α-Sooth Muscle Actin, respectively for evaluating the association of TF and microvascular properties. Pearson correlation was performed between percentage of TF expression area and DCE-MRI parameters, multiple microvascular indexes. Volume transfer constant (K Our results indicate that K