Merck
  • Isolation of a lectin binding rhamnogalacturonan-I containing pectic polysaccharide from pumpkin.
Isolation of a lectin binding rhamnogalacturonan-I containing pectic polysaccharide from pumpkin.
Carbohydrate polymers (2017-03-08)
Jing Zhao, Fuming Zhang, Xinyue Liu, Kalib St Ange, Anqiang Zhang, Quanhong Li, Robert J Linhardt
ZUSAMMENFASSUNG
A rhamnogalacturonan-I (RG-I) containing pectic polysaccharide (PPc) was isolated from pumpkin following a low-temperature alkali treatment and a combination of gradual alcohol precipitation and ion-exchange. Monosaccharide compositional analysis of PPc revealed the presence of rhamnose, galacturonic acid, galactose, and arabinose in a molar ratio of 7.4: 25: 28: 2.6. Structural and linkage analysis by 1D NMR (
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(±)-2-(p-Methoxyphenoxy)propionic acid, ≥98%