- Inhibitory effect of ethinylestradiol on coagulation factors in rats.
Inhibitory effect of ethinylestradiol on coagulation factors in rats.
Experimental animals (2016-11-11)
Yanira Franco-Murillo, Ruth Jaimez
PMID27829580
ZUSAMMENFASSUNG
Epidemiological and experimental data have indicated the beneficial and adverse effects of estrogenic replacement therapy. In the present study, we explored the effect of ethinylestradiol (EE) and 17β-estradiol (E
MATERIALIEN