Merck
  • Changes in expression of inhibitory substances in the intramural neurons of the stomach following streptozotocin- induced diabetes in the pig.
Changes in expression of inhibitory substances in the intramural neurons of the stomach following streptozotocin- induced diabetes in the pig.
World journal of gastroenterology (2017-10-04)
Michał Bulc, Katarzyna Palus, Łukasz Zielonka, Magdalena Gajęcka, Jarosław Całka
ZUSAMMENFASSUNG
Influence of chronic hyperglycemia on chemical coding of enteric neurons in stomach using pig as a model for human diabetic complications. Ten pigs were divided into two groups: diabetic (D group, In the control group in the myenteric ganglia (MG) of the corpus we have noted 22.28% ± 1.19% of nNOS positive neurons, while in diabetic group we have found 40.74% ± 2.22% of nNOS immunoreactive perikarya (increase by 82.85 %). In turn in the pylorus we have observed 15.91% ± 0.58% nNOS containing neurons in control animals and 35.38% ± 1.54% in the diabetes group (increase by 122.37%). In the MG of the antrum and submucosal ganglion (SG) in the corpus hyperglycemia did not cause statistically significant changes. With regard to VIP-positive cell bodies in the antrum MG in the control animals we have noted 18.38 ± 1.39% and 40.74% ± 1.77% in the experimental group (increase by 121.65%). While in the corpus we have observed 23.20% ± 0.23% in the control and 30.93% ± 0.86% in the diabetes group (increase by 33.31%). In turn in the pylorus VIP positive cells bodies constituted 23.64% ± 1.56% in the control group and 31.20% ± 1.10% in the experimental group (increase by 31.97%). In the submucosal ganglion in the corpus we have noted 43.61% ± 1.06% in the control animals and 37.00% ± 1.77% in the experimental group (decrease by 15.15%). Expression of GAL-positive perikarya showed statistically significant changes only in the MG of the antrum and pylorus. In the antrum GAL positive perykarya constituted 26.53% ± 1.52% in the control and 36.67% ± 1.02% in the experimental animals (increase by 38.22%). While in the pylorus GAL positive neurons in the control group constituted 16.32% ± 0.92% and 17.99% ± 0.38% in the experimental animals (increase by 10.23%). Our results support the hypothesis that in the course of diabetes, long term episodes of high glucose serum level may influence the chemical phenotyping of enteric neurons.