Merck
  • Endoglin controls blood vessel diameter through endothelial cell shape changes in response to haemodynamic cues.
Endoglin controls blood vessel diameter through endothelial cell shape changes in response to haemodynamic cues.
Nature cell biology (2017-05-23)
Wade W Sugden, Robert Meissner, Tinri Aegerter-Wilmsen, Roman Tsaryk, Elvin V Leonard, Jeroen Bussmann, Mailin J Hamm, Wiebke Herzog, Yi Jin, Lars Jakobsson, Cornelia Denz, Arndt F Siekmann
ZUSAMMENFASSUNG
The hierarchical organization of properly sized blood vessels ensures the correct distribution of blood to all organs of the body, and is controlled via haemodynamic cues. In current concepts, an endothelium-dependent shear stress set point causes blood vessel enlargement in response to higher flow rates, while lower flow would lead to blood vessel narrowing, thereby establishing homeostasis. We show that during zebrafish embryonic development increases in flow, after an initial expansion of blood vessel diameters, eventually lead to vessel contraction. This is mediated via endothelial cell shape changes. We identify the transforming growth factor beta co-receptor endoglin as an important player in this process. Endoglin mutant cells and blood vessels continue to enlarge in response to flow increases, thus exacerbating pre-existing embryonic arterial-venous shunts. Together, our data suggest that cell shape changes in response to biophysical cues act as an underlying principle allowing for the ordered patterning of tubular organs.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-α-Tubulin-Antikörper, monoklonaler Antikörper der Maus gegen, clone DM1A, purified from hybridoma cell culture
Sigma-Aldrich
Nifedipin, ≥98% (HPLC), powder
Sigma-Aldrich
E-64d, protease inhibitor