Merck
  • Mitogen-activated protein kinases are involved in hepatocanalicular dysfunction and cholestasis induced by oxidative stress.
Mitogen-activated protein kinases are involved in hepatocanalicular dysfunction and cholestasis induced by oxidative stress.
Archives of toxicology (2016-12-04)
Flavia D Toledo, Cecilia L Basiglio, Ismael R Barosso, Andrea C Boaglio, Andrés E Zucchetti, Enrique J Sánchez Pozzi, Marcelo G Roma
ZUSAMMENFASSUNG
In previous studies, we showed that the pro-oxidant model agent tert-butyl hydroperoxide (tBuOOH) induces alterations in hepatocanalicular secretory function by activating Ca
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Natriumtaurocholat, BRP, European Pharmacopoeia (EP) Reference Standard