Merck
CLEC5A is a critical receptor in innate immunity against Listeria infection.
Nature communications (2017-08-22)
Szu-Ting Chen, Fei-Ju Li, Tzy-Yun Hsu, Shu-Mei Liang, Yi-Chen Yeh, Wen-Yu Liao, Teh-Ying Chou, Nien-Jun Chen, Michael Hsiao, Wen-Bin Yang, Shie-Liang Hsieh
ZUSAMMENFASSUNG
The C-type lectin member 5A (CLEC5A) is a pattern recognition receptor for members of the Flavivirus family and has critical functions in response to dengue virus and Japanese encephalitis virus. Here we show that CLEC5A is involved in neutrophil extracellular trap formation and the production of reactive oxygen species and proinflammatory cytokines in response to Listeria monocytogenes. Inoculation of Clec5a
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Collagenase aus Clostridium histolyticum, suitable for release of physiologically active rat hepatocytes, Type IV, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
Deoxyribonuclease I aus Rinderpankreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Anti-Histone H3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution