- Systems analysis of ethanol production in the genetically engineered cyanobacterium
Systems analysis of ethanol production in the genetically engineered cyanobacterium
Biotechnology for biofuels (2017-03-14)
Joachim Kopka, Stefanie Schmidt, Frederik Dethloff, Nadin Pade, Susanne Berendt, Marco Schottkowski, Nico Martin, Ulf Dühring, Ekaterina Kuchmina, Heike Enke, Dan Kramer, Annegret Wilde, Martin Hagemann, Alexandra Friedrich
PMID28286551
ZUSAMMENFASSUNG
Future sustainable energy production can be achieved using mass cultures of photoautotrophic microorganisms, which are engineered to synthesize valuable products directly from CO Here, we performed a systems analysis of an engineered ethanol-producing strain of the cyanobacterium Systems analysis of ethanol production in
MATERIALIEN