Merck
  • Phorbol esters dPPA/dPA promote furin expression involving transcription factor CEBPβ in neuronal cells.
Phorbol esters dPPA/dPA promote furin expression involving transcription factor CEBPβ in neuronal cells.
Oncotarget (2017-09-28)
Jing-Si Zha, Bing-Lin Zhu, Lu Liu, Yu-Jie Lai, Yan Long, Xiao-Tong Hu, Xiao-Juan Deng, Xue-Feng Wang, Zhen Yan, Guo-Jun Chen
ZUSAMMENFASSUNG
Using high-throughput small molecule screening targeting furin gene, we identified that phorbol esters dPPA (12-Deoxyphorbol 13-phenylacetate 20-acetate) and dPA (12-Deoxyphorbol 13-acetate) significantly increased furin protein and mRNA expression in SH-SY5Y cells. This effect was prevented by PKC (protein kinase C) inhibitor calphostin C but not Ro318220, suggesting that the C1 domain, rather than the catalytic domain of PKC plays an important role. Luciferase assay revealed that nucleotides -7925 to -7426 were sufficient to mediate dPPA/dPA enhancement of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Phorbol-12,13-dibutyrat, ≥98% (TLC), powder
Sigma-Aldrich
U0126 ethanolate, ≥98% (HPLC), powder
Sigma-Aldrich
dPPA, ≥98% (HPLC)