- Effect of entinostat on NK cell-mediated cytotoxicity against osteosarcoma cells and osteosarcoma lung metastasis.
Effect of entinostat on NK cell-mediated cytotoxicity against osteosarcoma cells and osteosarcoma lung metastasis.
Oncoimmunology (2017-09-19)
Simin Kiany, Gangxiong Huang, Eugenie S Kleinerman
PMID28919994
ZUSAMMENFASSUNG
There is a crucial need for a new therapeutic approach for osteosarcoma (OS) lung metastasis since this disease remains the main cause of mortality in OS. We previously demonstrated that natural killer (NK) cell therapy has minimal efficacy against OS metastasis. This study determined whether the histone deacetylase inhibitor entinostat could immunosensitize OS cells to NK cell lysis and increases the efficacy of NK cell therapy for OS lung metastasis. Entinostat upregulated ligands for NK cell-activating receptors (major histocompatibility complex [MHC] class I polypeptide-related chain A [MICA] and B [MICB]; UL16 binding proteins 1, 2, 5, and 6; and CD155) on OS cells both