Merck
  • Gain-of-function p53 activates multiple signaling pathways to induce oncogenicity in lung cancer cells.
Gain-of-function p53 activates multiple signaling pathways to induce oncogenicity in lung cancer cells.
Molecular oncology (2017-04-20)
Catherine A Vaughan, Shilpa Singh, Steven R Grossman, Brad Windle, Swati Palit Deb, Sumitra Deb
ZUSAMMENFASSUNG
Gain-of-function (GOF) mutants of p53 upregulate genes implicated in cell proliferation and oncogenesis. Here, we report that GOF p53 induces tumorigenicity through simultaneous activation of key oncogenic pathways including those controlling putative tumor-initiating cell functions. We determined that in cells expressing p53-R273H, GOF p53 simultaneously upregulates genes from multiple signaling pathways by recognizing promoters containing distinct transcription factor (TF) binding sites. Our analytical data support a model in which GOF p53 complexes with two TFs on the promoter-a mediator protein, Med17, and a histone acetyl transferase, activating histone acetylation-and enhances gene expression to signal cell proliferation and oncogenesis. Thus, therapeutic inhibition of one GOF p53-induced pathway would be insufficient to prevent tumor growth as the oncoprotein activates a multitude of parallel pathways. This discovery suggests enormous selection advantage for cancer cells with GOF p53 to induce oncogenic growth, highlighting the problems of cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-γ-Tubulin-Antikörper, monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, clone GTU-88, purified from hybridoma cell culture
Sigma-Aldrich
Anti-NFκB p52-Antikörper, Upstate®, from mouse
Sigma-Aldrich
ChIPAb+ Acetyl-Histone H3 - ChIP Validated Antibody and Primer Set, from rabbit