Merck
  • On the use of ultracentrifugal devices for routine sample preparation in biomolecular magic-angle-spinning NMR.
On the use of ultracentrifugal devices for routine sample preparation in biomolecular magic-angle-spinning NMR.
Journal of biomolecular NMR (2017-02-24)
Abhishek Mandal, Jennifer C Boatz, Travis B Wheeler, Patrick C A van der Wel
ZUSAMMENFASSUNG
A number of recent advances in the field of magic-angle-spinning (MAS) solid-state NMR have enabled its application to a range of biological systems of ever increasing complexity. To retain biological relevance, these samples are increasingly studied in a hydrated state. At the same time, experimental feasibility requires the sample preparation process to attain a high sample concentration within the final MAS rotor. We discuss these considerations, and how they have led to a number of different approaches to MAS NMR sample preparation. We describe our experience of how custom-made (or commercially available) ultracentrifugal devices can facilitate a simple, fast and reliable sample preparation process. A number of groups have since adopted such tools, in some cases to prepare samples for sedimentation-style MAS NMR experiments. Here we argue for a more widespread adoption of their use for routine MAS NMR sample preparation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allylmethylsulfon, 96%