Merck
ZnO nanoparticles sensitized by CuInZn
Beilstein journal of nanotechnology (2017-07-08)
Florian Donat, Serge Corbel, Halima Alem, Steve Pontvianne, Lavinia Balan, Ghouti Medjahdi, Raphaël Schneider
ZUSAMMENFASSUNG
Alloyed CuInZn