Merck
A new neolignan from Selaginella moellendorffii Hieron.
Natural product research (2017-03-09)
Wei Zeng, Cai-Ping Yao, Ping-Sheng Xu, Guo-Gang Zhang, Zhao-Qian Liu, Kang-Ping Xu, Zhen-Xing Zou, Gui-Shan Tan
ZUSAMMENFASSUNG
A new neolignan, selamoellenin A (1), was isolated from the whole plants of Selaginella moellendorffii Hieron. The structure was elucidated on the basis of comprehensive spectroscopic methods (1D/2D NMR and HRMS). Compound 1 was evaluated for its protective effect against high glucose-induced human umbilical vein endothelial cells (HUVECs) damage in vitro.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%