Merck
A dual-layer macro/mesoporous structured TiO
Materials science & engineering. C, Materials for biological applications (2017-06-04)
Ranran Zhang, Tarek A Elkhooly, Qianli Huang, Xujie Liu, Xing Yang, Hao Yan, Zhiyuan Xiong, Jing Ma, Qingling Feng, Zhijian Shen
ZUSAMMENFASSUNG
A dual-layer TiO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
2-Phenylindol, technical grade, 95%