Merck
  • Flavanol Quantification of Grapes via Multiple Reaction Monitoring Mass Spectrometry. Application to Differentiation among Clones of Vitis vinifera L. cv. Rufete Grapes.
Flavanol Quantification of Grapes via Multiple Reaction Monitoring Mass Spectrometry. Application to Differentiation among Clones of Vitis vinifera L. cv. Rufete Grapes.
Journal of agricultural and food chemistry (2017-02-06)
Ignacio García-Estévez, Cristina Alcalde-Eon, M Teresa Escribano-Bailón
ZUSAMMENFASSUNG
The determination of the detailed flavanol composition in food matrices is not a simple task because of the structural similarities of monomers and, consequently, oligomers and polymers. The aim of this study was the development and validation of an HPLC-MS/MS-multiple reaction monitoring (MRM) method that would allow the accurate and precise quantification of catechins, gallocatechins, and oligomeric proanthocyanidins. The high correlation coefficients of the calibration curves (>0.993), the recoveries not statistically different from 100%, the good intra- and interday precisions (<5%), and the LOD and LOQ values, low enough to quantify flavanols in grapes, are good results from the method validation procedure. Its usefulness has also been tested by determining the detailed composition of Vitis vinifera L. cv. Rufete grapes. Seventy-two (38 nongalloylated and 34 galloylated) and 53 (24 procyanidins and 29 prodelphinidins) flavanols have been identified and quantified in grape seed and grape skin, respectively. The use of HCA and PCA on the detailed flavanol composition has allowed differentiation among Rufete clones.