- Inhibition of MHC-I by Brucella abortus is an early event during infection and involves EGFR pathway.
Inhibition of MHC-I by Brucella abortus is an early event during infection and involves EGFR pathway.
Immunology and cell biology (2016-11-05)
Lis N Velásquez, M Ayelén Milillo, M Victoria Delpino, Aldana Trotta, M Florencia Mercogliano, Roberto G Pozner, Roxana Schillaci, Patricia V Elizalde, Guillermo H Giambartolomei, Paula Barrionuevo
PMID27811842
ZUSAMMENFASSUNG
Brucella abortus is able to persist inside the host despite the development of potent CD8
MATERIALIEN