Merck
Carbon-Nanodot Solar Cells from Renewable Precursors.
ChemSusChem (2017-01-21)
Adam Marinovic, Lim S Kiat, Steve Dunn, Maria-Magdalena Titirici, Joe Briscoe
ZUSAMMENFASSUNG
It has recently been shown that waste biomass can be converted into a wide range of functional materials, including those with desirable optical and electronic properties, offering the opportunity to find new uses for these renewable resources. Photovoltaics is one area in which finding the combination of abundant, low-cost and non-toxic materials with the necessary functionality can be challenging. In this paper the performance of carbon nanodots derived from a wide range of biomaterials obtained from different biomass sources as sensitisers for TiO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Cystein, technical grade