Merck
Halide perovskite solar cells using monocrystalline TiO
Nanotechnology (2017-05-31)
Ujwal Kumar Thakur, Abdelrahman M Askar, Ryan Kisslinger, Benjamin D Wiltshire, Piyush Kar, Karthik Shankar
ZUSAMMENFASSUNG
This is the first report of a 17.6% champion efficiency solar cell architecture comprising monocrystalline TiO
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-tert-Butylpyridin, 98%
Sigma-Aldrich
Formamidiniumiodid