Merck
  • Four new steroids from the endophytic fungus Chaetomium sp. M453 derived of Chinese herbal medicine Huperzia serrata.
Four new steroids from the endophytic fungus Chaetomium sp. M453 derived of Chinese herbal medicine Huperzia serrata.
Fitoterapia (2017-01-04)
Fei-Xue Yu, Zhe Li, Yao Chen, Yin-He Yang, Guo-Hong Li, Pei-Ji Zhao
ZUSAMMENFASSUNG
An endophytic fungus, Chaetomium sp. M453, was isolated from Huperzia serrata (Thunb. ex Murray) Trev. and subjected to phytochemical investigation. Three unusual C25 steroids, neocyclocitrinols E-G (1-3), and 3β-hydroxy-5,9-epoxy-(22E,24R)-ergosta-7,22-dien-6-one (4) together with three known steroids were isolated from solid fermentation products of the fungus, which were elucidated by extensive spectroscopic analyses, including 1D-, 2D-NMR, and HR-ESI-MS experiments. The absolute configuration of 1 was determined by X-ray crystallographic analysis and CD analyses. The acetylcholinesterase inhibitory activities of compounds 1-4 were tested in vitro. Compound 4 showed weak acetylcholinesterase inhibitory activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,3-Propylene sulfite, 99%