Merck
  • MicroRNA-214 modulates neural progenitor cell differentiation by targeting Quaking during cerebral cortex development.
MicroRNA-214 modulates neural progenitor cell differentiation by targeting Quaking during cerebral cortex development.
Scientific reports (2017-08-16)
Pengcheng Shu, Hongye Fu, Xiangyu Zhao, Chao Wu, Xiangbin Ruan, Yi Zeng, Wei Liu, Ming Wang, Lin Hou, Pan Chen, Bin Yin, Jiangang Yuan, Boqin Qiang, Xiaozhong Peng
ZUSAMMENFASSUNG
The accurate generation of an appropriate number of different neuronal and glial subtypes is fundamental to normal brain functions and requires tightly orchestrated spatial and temporal developmental programmes to maintain the balance between the proliferation and the differentiation of neural progenitor cells. However, the molecular mechanism governing this process has not been fully elucidated. Here, we found that miR-214-3p was highly expressed in neural progenitor cells and dynamically regulated during neocortical development. Moreover, our in vivo and in vitro studies showed that miR-214 inhibited self-renewal of neural progenitor cells and promoted neurogenesis. In addition, after target screening, we identified miR-214 targets including Quaking (Qki) by binding the 3'- untranslated region (3'-UTR) of the Qki mRNA, which was specifically expressed in the progenitor cells of the proliferative ventricular zone as 3 Qki isoforms. Furthermore, overexpression and knockdown of Qki showed that the different isoforms of Qki had different functions in the regulation of neural progenitor cells differentiation. Moreover, overexpression of Qki could counteract the function of miR-214 in neurogenesis. Our results revealed that miR-214 maintains the balance between neural progenitor/stem cell proliferation and differentiation together with Quaking, its target gene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
L-(−)-Glukose, ≥99%