Merck
  • Inhibitory effects of fifteen phthalate esters in human cDNA-expressed UDP-glucuronosyltransferase supersomes.
Inhibitory effects of fifteen phthalate esters in human cDNA-expressed UDP-glucuronosyltransferase supersomes.
Chemosphere (2017-07-30)
Yun-Feng Cao, Zuo Du, Zhi-Tu Zhu, Hong-Zhi Sun, Zhi-Wei Fu, Kun Yang, Yong-Zhe Liu, Cui-Min Hu, Pei-Pei Dong, Frank J Gonzalez, Zhong-Ze Fang
ZUSAMMENFASSUNG
Phthalate esters (PAEs) have been extensively used in industry as plasticizers and there remains concerns about their safety. The present study aimed to determine the inhibition of phthalate esters (PAEs) on the activity of the phase II drug-metabolizing enzymes UDP-glucuronosyltransferases (UGTs). In vitro recombinant UGTs-catalyzed glucuronidation of 4-methylumbelliferone was used to investigate the inhibition potentials of PAEs towards various s UGTs. PAEs exhibited no significant inhibition of UGT1A1, UGT1A3, UGT1A8, UGT1A10, UGT2B15, and UGT2B17, and limited inhibition of UGT1A6, UGT1A7 and UGT2B4. However, UGT1A9 was strongly inhibited by PAEs. In silico docking demonstrated a significant contribution of hydrogen bonds and hydrophobic interactions contributing to the inhibition of UGT by PAEs. The K
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Uridin-5′-diphosphoglucuronsäure Ammoniumsalz, 98-100%