Merck
  • The higher the better? Differences in phenolics and cyanogenic glycosides in Sambucus nigra leaves, flowers and berries from different altitudes.
The higher the better? Differences in phenolics and cyanogenic glycosides in Sambucus nigra leaves, flowers and berries from different altitudes.
Journal of the science of food and agriculture (2016-10-14)
Mateja Senica, Franci Stampar, Robert Veberic, Maja Mikulic-Petkovsek
ZUSAMMENFASSUNG
Elderberry (Sambucus nigra L.) possesses high antioxidant activity and has been used to treat numerous medicinal disorders. In addition to their antioxidant properties, elderberry parts accumulate toxic cyanogenic glycosides (CGG). It has been proven that altitude influences the biosynthesis of many secondary metabolites. In the present study we investigated the change of phenolics and CGG in elder leaves, flowers, and berries induced by different altitudes and locations. The data indicate that the accumulation of CGG and phenolics is affected by the altitude of the growing site. An increase of anthocyanin content was recorded in elder berries collected at higher elevations in both locations. Fruit collected at the foothills of location 2 contained 3343 µg g Elder berries and flowers collected at the foothill were characterised by the lowest levels of both beneficial (phenolics) and harmful compounds (CGG) and are suitable for moderate consumption. © 2016 Society of Chemical Industry.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Prunasin, ≥90% (LC/MS-ELSD)