Merck
  • Mapping Atheroprotective Functions and Related Proteins/Lipoproteins in Size Fractionated Human Plasma.
Mapping Atheroprotective Functions and Related Proteins/Lipoproteins in Size Fractionated Human Plasma.
Molecular & cellular proteomics : MCP (2017-02-23)
Debi K Swertfeger, Hailong Li, Sandra Rebholz, Xiaoting Zhu, Amy S Shah, W Sean Davidson, Long J Lu
ZUSAMMENFASSUNG
HDL has been shown to possess a variety of cardio-protective functions, including removal of excess cholesterol from the periphery, and inhibition of lipoprotein oxidation. It has been proposed that various HDL subparticles exist, each with distinct protein and lipid compositions, which may be responsible for HDL's many functions. We hypothesized that HDL functions will co-migrate with the operational lipoprotein subspecies when separated by gel filtration chromatography. Plasma from 10 healthy male donors was fractionated and the protein composition of the phospholipid containing fractions was analyzed by mass spectrometry (MS). Each fraction was evaluated for its proteomic content as well as its ability to promote cholesterol efflux and protect low density lipoprotein (LDL) from free radical oxidation. For each function, several peaks of activity were identified across the plasma size gradient. Neither cholesterol efflux or LDL antioxidation activity correlated strongly with any single protein across the fractions. However, we identified multiple proteins that had strong correlations (r values >0.7,
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zyklisches 8-Bromadenosin-3′,5′-Monophosphat Natriumsalz, ≥97% (HPLC), powder