Merck
Efficient synthesis of secondary amines by reductive amination of curdlan Staudinger ylides.
Carbohydrate polymers (2017-06-06)
Ruoran Zhang, Shu Liu, Kevin J Edgar
ZUSAMMENFASSUNG
Staudinger-related reactions between azides and phosphines are important in organic chemistry due to their chemoselectivity, high efficiency, and mild reaction conditions. Staudinger reduction of azides affords highly reactive iminophosphorane ylides; the reactivity of the negatively charged ylide nitrogen atom has not previously been fully explored in polysaccharide chemistry. Curdlan, a natural, biocompatible, and bioactive β-1,3-glucan with low toxicity, has remarkable potential in biomedical and pharmaceutical applications. Herein we describe a new method for preparation of regioselectively iminated/aminated curdlan derivatives via a Staudinger ylide. 6-Azido-6-deoxy-2,4-di-O-acyl-curdlan was treated with triphenylphosphine to generate the highly nucleophilic iminophosphorane intermediate which afforded: i) 6-imino curdlans by reaction with several aromatic aldehydes, and ii) 6-monoalkylamino curdlans by reductive amination using these aldehydes and NaBH
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2-Pyridincarbaldehyd, 99%
Sigma-Aldrich
4-Chlorbenzaldehyd, 97%