Merck
LMTK3 knockdown retards cell growth and invasion and promotes apoptosis in thyroid cancer.
Molecular medicine reports (2017-03-06)
Lu Lu, Xueli Yuan, Qiang Zhang, Hong Zhang, Baozhong Shen
ZUSAMMENFASSUNG
Lemur tyrosine kinase-3 (LMTK3) is a member of the serine/threonine tyrosine kinase family, which is thought to be involved in tumor progression and prognosis. The purpose of the present study was to determine the diagnostic significance and therapeutic targets in thyroid cancer. ELISA assay was used to detect the protein expression of serum LMTK3. Immunohistochemistry and reverse transcription‑quantitative polymerase chain reaction were employed to measure the expression of LMTK3. Flow cytometry was used to determine the cell cycle. Transwell assay was used to measure the invasion and migration of SW579 cells and terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) assay to detect cell apoptosis. The LMTK3 level was positively associated with disease stage and pathological type, whereas there was no correlation between LMTK3 level and gender, age, tumor size or lymph node metastasis. The serum LMTK3 level was significantly increased in 102 thyroid carcinoma patients compared with 52 benign thyroid tumor patients and 50 healthy volunteers (P=0.001). The protein and mRNA expression of LMTK3 was markedly higher in thyroid cancer patients compared with patients with benign thyroid tumors. Notably, LMTK3 knockdown retarded proliferation, invasion and migration in SW579 cells. In addition, downregulation of LMTK3 promoted apoptosis in SW579 cells. These findings indicated that LMTK3 knockdown retards the growth of thyroid cancer cells partly through inhibiting proliferation, invasion, migration and inducing apoptosis in SW579 cells. It may serve as a useful diagnostic biomarker and a novel therapeutic target for patients with thyroid cancer.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human LMTK3