- Study on dose-dependent, frequency-dependent, and accumulative effects of 1.5 GHz and 2.856 GHz microwave on cognitive functions in Wistar rats.
Study on dose-dependent, frequency-dependent, and accumulative effects of 1.5 GHz and 2.856 GHz microwave on cognitive functions in Wistar rats.
Scientific reports (2017-09-09)
Shengzhi Tan, Hui Wang, Xinping Xu, Li Zhao, Jing Zhang, Ji Dong, Binwei Yao, Haoyu Wang, Hongmei Zhou, Yabing Gao, Ruiyun Peng
PMID28883530
ZUSAMMENFASSUNG
Many studies have revealed the cognitive decline induced by microwave radiation. However, the systematic study on dose-dependent, frequency-dependent and accumulative effects of microwave exposure at different frequencies was lacking. Here, we studied the relationship between the effects and the power and frequency of microwave and analyzed the accumulative effects of two different frequency microwaves with the same average power density. After microwave radiation, declines in spatial learning and memory and fluctuations of brain electric activities were found in the 10 mW/cm