Merck
N
Chemical research in toxicology (2017-07-12)
Bahar Edrissi, Koli Taghizadeh, Benjamin C Moeller, Rui Yu, Dean Kracko, Melanie Doyle-Eisele, James A Swenberg, Peter C Dedon
ZUSAMMENFASSUNG
Exposure to both endogenous and exogenous formaldehyde has been established to be carcinogenic, likely by virtue of forming nucleic acid and proteins adducts such as N