Merck
  • Calreticulin modulates the intracellular survival of mycobacteria by regulating ER-stress-mediated apoptosis.
Calreticulin modulates the intracellular survival of mycobacteria by regulating ER-stress-mediated apoptosis.
Oncotarget (2017-09-25)
Sung Hee Jo, Ji-Ae Choi, Yun-Ji Lim, Junghwan Lee, Soo-Na Cho, Sung-Man Oh, Dam Go, Seon-Hwa Kim, Chang-Hwa Song
ZUSAMMENFASSUNG
Endoplasmic reticulum (ER)-stress-mediated apoptosis is a host defense mechanism against
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)