- Hepatoprotective and antioxidant activity of quercetin loaded chitosan/alginate particles in vitro and in vivo in a model of paracetamol-induced toxicity.
Hepatoprotective and antioxidant activity of quercetin loaded chitosan/alginate particles in vitro and in vivo in a model of paracetamol-induced toxicity.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-06-04)
Virginia Tzankova, Denitsa Aluani, Magdalena Kondeva-Burdina, Yordan Yordanov, Feodor Odzhakov, Alexandar Apostolov, Krassimira Yoncheva
PMID28577496
ZUSAMMENFASSUNG
The toxic liver impairment caused by free radical injury or excessive reactive oxigen species (ROS) formation could be effectivelly attenuated by natural antioxidants. The present study aimed to explore and compare the hepatoprotective and antioxidant effects of free and encapsulated quercetin in in vitro and in vivo models of hepatotoxicity. Thus, quercetin was encapsulated in chitosan/alginate nanoparticles by gelation method. Both empty and quercetin-loaded nanoparticles revealed good safety profile in vitro, determined by the lack of cytotoxicity in human hepatoma HepG2 cells. The pretreatment of HepG2 cells with encapsulated quercetin (10μg/ml) significantly attenuated the decrease in cell viability in H