Merck
  • Determining the distribution of pharmaceutically active compounds (PhACs) in soils and sediments by pressurized hot water extraction (PHWE).
Determining the distribution of pharmaceutically active compounds (PhACs) in soils and sediments by pressurized hot water extraction (PHWE).
Chemosphere (2017-07-30)
Miriam Biel-Maeso, Carmen Corada-Fernández, Pablo A Lara-Martín
ZUSAMMENFASSUNG
The occurrence of pharmaceutically active compounds (PhACs) in the environment may pose a potential risk for humans and ecosystems. Wastewater treatment plants (WWTPs) are recognized as one of the main sources of these chemicals into both aquatic and terrestrial environments. The objectives of the study were to determine the presence of a wide variety of PhACs (n = 45) in sewage-impacted sediments and soils from the Guadalete River basin (SW Spain) by developing and applying an environmentally friendly multi-residue method based on pressurized hot water extraction (PHWE). Different parameters were optimized, including extraction temperature, pH, solvents, and clean-up. Extraction recoveries were analyte dependent, varying between 50% and 140% for most of the analytes when using pure water as extraction solvent at 100 °C. Determination of PhACs was performed by ultra-high performance liquid chromatography - tandem mass spectrometry (UPLC-MS/MS), enabling method detection (mLODs) and quantification (mLOQs) limits between <0.01 and 0.83 ng g
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Sulfadimethoxin-d6, VETRANAL®, analytical standard
Supelco
Ofloxacin-d3, VETRANAL®, analytical standard