Merck
  • A novel and efficient fungal delignification strategy based on versatile peroxidase for lignocellulose bioconversion.
A novel and efficient fungal delignification strategy based on versatile peroxidase for lignocellulose bioconversion.
Biotechnology for biofuels (2017-09-20)
Wen Kong, Xiao Fu, Lei Wang, Ahmad Alhujaily, Jingli Zhang, Fuying Ma, Xiaoyu Zhang, Hongbo Yu
ZUSAMMENFASSUNG
The selective lignin-degrading white-rot fungi are regarded to be the best lignin degraders and have been widely used for reducing the saccharification recalcitrance of lignocellulose. However, the biological delignification and conversion of lignocellulose in biorefinery is still limited. It is necessary to develop novel and more efficient bio-delignification systems. From the highly efficient system of enzymatic recalcitrance removal by new white-rot fungus, we identified a new delignification strategy based on VP which could oxidize both phenolic and nonphenolic lignin units and break different linkages in lignin. In addition, this is the first evidence that VP could break 5-5' linkage efficiently in vitro. Moreover, VP improved the enzymatic hydrolysis of corn stover in vitro. The remarkable lignin-degradative potential makes VP attractive for biotechnological applications.