Merck
  • The PTEN inhibitor bpV(pic) promotes neuroprotection against amyloid β-peptide (25-35)-induced oxidative stress and neurotoxicity.
The PTEN inhibitor bpV(pic) promotes neuroprotection against amyloid β-peptide (25-35)-induced oxidative stress and neurotoxicity.
Neurological research (2017-04-25)
Xiao-Ying Liu, Li-Jing Zhang, Zhou Chen, Li-Bin Liu
ZUSAMMENFASSUNG
The aim of this study was to elucidate the mechanism underlying the neuroprotective effects of the phosphatase and tensin homolog (PTEN) inhibitor, bisperoxovanadium-pic [bpV(pic)]. We determined the effects of bpV(pic) on amyloid-β-peptide-(25-35)-induced neurotoxicity, particularly intracellular reactive oxygen species (ROS) production and mitochondria-mediated apoptotic signaling, in a human neuroblastoma (SH-SY5Y) cell model. We found that exposure of SH-SY5Y cells to amyloid β peptides (Aβ Aβ peptides are thought to cause neurodegeneration in Alzheimer's disease (AD), via the induction of free radical oxidative stress. Our results indicate that bpV(pic) provides protection against Aβ
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)