- Lower levels of vitamin A are associated with increased gastrointestinal graft-versus-host disease in children.
Lower levels of vitamin A are associated with increased gastrointestinal graft-versus-host disease in children.
Blood (2017-03-11)
Dana T Lounder, Pooja Khandelwal, Christopher E Dandoy, Sonata Jodele, Michael S Grimley, Gregory Wallace, Adam Lane, Cynthia Taggart, Ashley C Teusink-Cross, Kelly E Lake, Stella M Davies
PMID28279965
ZUSAMMENFASSUNG
Vitamin A promotes development of mucosal tolerance and enhances differentiation of regulatory T cells. Vitamin A deficiency impairs epithelial integrity, increasing intestinal permeability. We hypothesized that higher vitamin A levels would reduce the risk of graft-versus-host disease (GVHD) through reduced gastrointestinal (GI) permeability, reduced mucosal injury, and reduced lymphocyte homing to the gut. We tested this hypothesis in a cohort study of 114 consecutive patients undergoing allogeneic stem cell transplant. Free vitamin A levels were measured in plasma at day 30 posttransplant. GI GVHD was increased in patients with vitamin A levels below the median (38% vs 12.4% at 100 days,