- Nortirucallane A, a new tirucallane-type nortriterpenoid isolated from Lonicerae japonicae flos.
Nortirucallane A, a new tirucallane-type nortriterpenoid isolated from Lonicerae japonicae flos.
Natural product research (2017-03-11)
Xu Wang, Jing Zhang, Ya-Qun Li, Bao-Xiang Du, Qi-Hui Sun, Yun-Jiao Cao, Hai-Qiang Jiang, Yu-Hong Liu, Hong-Lei Zhou, Rong Rong
PMID28281370
ZUSAMMENFASSUNG
One new tirucallane-type nortriterpenoid Nortirucallane A (1), together with Chrysoeriol (2) and Isorhamnetin (3), was isolated from the methylene chloride part of Lonicerae japonicae flos. Their structures were elucidated by the detailed analysis of comprehensive spectroscopic data. Compound 3 was isolated from the genus of Lonicera for the first time. The significance of 1 for the study of phytochemical taxonomy was discussed.
MATERIALIEN