Merck
  • Analysis of polyphenols in brewer's spent grain and its comparison with corn silage and cereal brans commonly used for animal nutrition.
Analysis of polyphenols in brewer's spent grain and its comparison with corn silage and cereal brans commonly used for animal nutrition.
Food chemistry (2017-09-07)
Flávia Santi Stefanello, Clarissa Obem Dos Santos, Vivian Caetano Bochi, Ana Paula Burin Fruet, Marcela Bromenberg Soquetta, Andréa Cristina Dörr, José Laerte Nörnberg
ZUSAMMENFASSUNG
Brewer's spent grain (BSG) could be tested as an alternative source of polyphenols in animal nutrition. Proper extraction and analytical methods are critical for quantification. Thus, extraction for BSG, corn silage, and brans of rice, corn, and wheat were studied for the highest yield of polyphenols. A method for 18 phenolic monomers by HPLC-DAD was developed, validated, and applied to samples. An aqueous solution of NaOH (0.75% w/v) using integral samples for extraction resulted in the highest values for colorimetric measurements in all analyzed sources. Method by maceration showed the highest phenolic yield when applied in corn silage and BSG. However, for brans the best method was microwave assisted. Results from HPLC-DAD analysis clearly showed that native structures of phenolic compounds were simplified to its monomers allowing quantification and sample discrimination. BSG had the highest concentration of polyphenols and could be a promising and innovative source for animal feed studies.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Quercetin, ≥95% (HPLC), solid
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
p-Coumarsäure, ≥98.0% (HPLC)
Sigma-Aldrich
Kaffeesäure, ≥98.0% (HPLC)
Sigma-Aldrich
Chlorogensäure, ≥95% (titration)
Sigma-Aldrich
Syringasäure, ≥95% (HPLC)
Sigma-Aldrich
Kaempferol, ≥90% (HPLC), powder