Merck
  • Tristetraprolin inhibits mitochondrial function through suppression of α-Synuclein expression in cancer cells.
Tristetraprolin inhibits mitochondrial function through suppression of α-Synuclein expression in cancer cells.
Oncotarget (2017-04-15)
Mai-Tram Vo, Seong Hee Choi, Ji-Heon Lee, Chung Hwan Hong, Jong Soo Kim, Unn Hwa Lee, Hyung-Min Chung, Byung Ju Lee, Jeong Woo Park, Wha Ja Cho
ZUSAMMENFASSUNG
Mitochondrial dynamics play critical roles in maintaining mitochondrial functions. Here, we report a novel mechanism for regulation of mitochondrial dynamics mediated by tristetraprolin (TTP), an AU-rich element (ARE)-binding protein. Overexpression of TTP resulted in elongated mitochondria, down-regulation of mitochondrial oxidative phosphorylation, reduced membrane potential, cytochrome c release, and increased apoptotic cell death in cancer cells. TTP overexpression inhibited the expression of α-Synuclein (α-Syn). TTP bound to the ARE within the mRNA 3'-untranslated regions (3'-UTRs) of α-Syn and enhanced the decay of α-Syn mRNA. Overexpression of α-Syn without the 3'-UTR restored TTP-induced defects in mitochondrial morphology, mitochondrial oxidative phosphorylation, membrane potential, and apoptotic cell death. Taken together, our data demonstrate that TTP acts as a regulator of mitochondrial dynamics through enhancing degradation of α-Syn mRNA in cancer cells. This finding will increase understanding of the molecular basis of mitochondrial dynamics.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
IgG aus Mausserum, reagent grade, ≥95% (SDS-PAGE), lyophilized powder
Sigma-Aldrich
Thymidine 5′-triphosphate sodium salt, ≥96%
Sigma-Aldrich
Tetramethylrhodamin-methylester-perchlorat, ≥95%